3-Bromopyrazolo[1,5-a]pyridine 97%
Supplier: Ambeed
|
Warning
|
A235054-250MG
A235054-1G
103810-990EA
228
USD
103810-990
103850-754
76987-490
77089-754
3-Bromopyrazolo[1,5-a]pyridine 97%
5-Bromopyrazolo[1,5-a]pyridine
Precaución: for Research Use Only
Formula:
C₇H₅BrN₂ MW: 197.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06661764 CAS Number: 5910-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...