3-Bromo-1-propanol 97%

Supplier: Ambeed
Danger

Synonyms: 3-Bromopropan-1-ol, 1-Bromo-3-hydroxypropane

A316010-1000G A316010-100G A316010-500G A316010-25G A316010-5G
103810-092EA 221 USD
103810-092 103836-014 76838-852 77277-616 77277-618 77306-140 77306-142
3-Bromo-1-propanol 97%
3-Bromo-1-propanol

Precaución: for Research Use Only

Formula: BrH₂CCH₂CH₂OH
MW: 138.99 g/mol
Boiling Pt: 62 °C (5 mm Hg)
Flash Pt: 101.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002942
CAS Number: 627-18-9
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR