3-Brompropylamine hydrobromide 95%

Supplier: Ambeed
Warning

A287264-1000G A287264-100G A287264-10G A287264-500G A287264-25G
103834-882EA 7 USD
103834-882 77277-096 77277-098 77277-100 77277-102 77305-512
3-Brompropylamine hydrobromide 95%
3-Brompropylamine hydrobromide

Precaución: for Research Use Only

Formula: BrH₂CCH₂CH₂NH₃Br
MW: 218.92 g/mol
Melting Pt: 171…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00012912
CAS Number: 5003-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR