3-Bromopiperidine-2,6-dione 97%

Supplier: Ambeed
Warning

A875928-1G A875928-5G A875928-250MG A875928-25G A875928-500G
103841-300EA 25 USD
103841-300 76845-566 76845-568 77171-726 77171-728 77893-676
3-Bromopiperidine-2,6-dione 97%
3-Bromopiperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₅H₆BrNO₂
Boiling Pt: 336.4 °C (760 mm Hg)
Flash Pt: 157.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11053059
CAS Number: 62595-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR