3-Bromophenylacetylene 97%
Supplier: Ambeed
|
Warning
|
A100389-100G
A100389-25G
A100389-5G
A100389-1G
103795-410EA
7
USD
103795-410
103821-108
77188-518
77188-520
77282-486
77296-662
3-Bromophenylacetylene 97%
3-Bromophenylacetylene
Precaución: for Research Use Only
Formula:
C₈H₅Br Boiling Pt: 211.8 °C (760 mm Hg) Flash Pt: 85.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03839983 CAS Number: 766-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...