3-Bromo-N-methylaniline 98%

Supplier: Ambeed
Warning

A229344-1G A229344-25G A229344-5G A229344-100G A229344-10G A229344-250MG
103819-464EA 7 USD
103819-464 103850-364 77089-192 77089-194 77089-196 77275-730 77275-732 77275-734
3-Bromo-N-methylaniline 98%
3-Bromo-N-methylaniline

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 253 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD05664376
CAS Number: 66584-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR