3-Bromo-N,N-dimethylpropan-1-amine hydrobromide 95%

Supplier: Ambeed
Warning

A124344-10G A124344-250MG A124344-25G A124344-1G A124344-5G
103800-520EA 17 USD
103800-520 77271-712 77271-714 77271-716 77297-986 77297-988
3-Bromo-N,N-dimethylpropan-1-amine hydrobromide 95%
3-Bromo-N,N-dimethylpropan-1-amine hydrobromide

Precaución: for Research Use Only

Formula: C5H13Br2N
MW: 246.9714 g/mol
Flash Pt: 246.9 °C
Storage Temperature: Ambient
MDL Number: MFCD11109991
CAS Number: 5845-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR