3-Bromo-N,N-dimethylaniline 96%

Supplier: Ambeed
Danger

A527342-1G A527342-25G A527342-5G A527342-10G
103828-544EA 9 USD
103828-544 103839-656 76843-666 76843-668 76843-670 77103-274
3-Bromo-N,N-dimethylaniline 96%
3-Bromo-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 259.7 °C (760 mm Hg)
Melting Pt: 11 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00045020
CAS Number: 16518-62-0
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR