(3-Bromopropyl)trimethylammonium bromide, (contains approx. 3% H₂O) 97% (contains ~3% H₂O)

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-N,N,N-trimethylpropan-1-aminium bromide

A446294-25G A446294-100G A446294-10G A446294-500G
103838-806EA 5 USD
103838-806 76842-600 77143-460 77143-462 77616-910
(3-Bromopropyl)trimethylammonium bromide, (contains approx. 3% H₂O) 97% (contains ~3% H₂O)
(3-Bromopropyl)trimethylammonium bromide

Precaución: for Research Use Only

Formula: C₆H₁₅Br₂N
Melting Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00075396
CAS Number: 3779-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR