3-Bromo-N-tert-butylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-N-(tert-butyl)benzenesulphonamide

A198998-250MG A198998-1G A198998-5G
103817-068EA 19 USD
103817-068 103847-692 77030-084 77085-726 77085-728
3-Bromo-N-tert-butylbenzenesulfonamide 98%
3-Bromo-N-tert-butylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrNO₂S
MW: 292.1900 g/mol
Boiling Pt: 365.3 °C (760 mm Hg)
Melting Pt: 68…70 °C at N/A mmHg
Flash Pt: 174.7 °C
Storage Temperature: Ambient
MDL Number: MFCD07363824
CAS Number: 308283-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR