3-(Bromomethyl)phenylboronic acid 97%

Supplier: Ambeed
Danger

Synonyms: 3-Bromomethylphenylboronic acid, 3-(Bromomethyl)benzeneboronic acid, Dihydroxy(3-(bromomethyl)phenyl)borane, (3-(Bromomethyl)phenyl)metaboric acid

A177091-25G A177091-5G A177091-10G A177091-1G
103807-210EA 10 USD
103807-210 103845-536 77082-914 77082-916 77191-598 77301-494
3-(Bromomethyl)phenylboronic acid 97%
3-(Bromomethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₂
Boiling Pt: 359.1 °C (760 mm Hg)
Melting Pt: 214…216 °C
Flash Pt: 171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01632207
CAS Number: 51323-43-4
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR