3-Bromoisoxazole 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-1,2-oxazole

A110194-100MG A110194-250MG
103797-638EA 48 USD
103797-638 103823-444 76796-770 77666-606
3-Bromoisoxazole 97%
3-Bromoisoxazole

Precaución: for Research Use Only

Formula: C₃H₂BrNO
MW: 147.958 g/mol
Boiling Pt: 182.7±13.0 °C (760 mm Hg)
Flash Pt: 64.3 °C
Storage Temperature: Freezer
MDL Number: MFCD07778379
CAS Number: 111454-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR