3-Bromoisoquinoline 98%
Supplier: Ambeed
|
Warning
|
A162934-100MG
A162934-10G
A162934-250MG
A162934-1G
A162934-25G
A162934-5G
103805-672EA
18
USD
103805-672
103843-930
77273-410
77273-412
77273-414
77320-002
77320-004
77320-006
3-Bromoisoquinoline 98%
3-Bromoisoquinoline
Precaución: for Research Use Only
Formula:
C₉H₆BrN Boiling Pt: 316.3 °C (760 mm Hg) Melting Pt: 60…64 °C Flash Pt: 145.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234479 CAS Number: 34784-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...