3-Bromoisonicotinic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromopyridine-4-carboxylic acid, 3-Bromo-4-pyridinecarboxylic acid
A258659-25G
A258659-5G
A258659-100G
A258659-10G
A258659-1G
103812-392EA
8
USD
103812-392
103852-268
77091-804
77091-806
77289-160
77289-162
77304-746
3-Bromoisonicotinic acid 98%
3-Bromoisonicotinic acid
Precaución: for Research Use Only
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol Melting Pt: 234…238 °C Flash Pt: 235 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040944 CAS Number: 13959-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...