3-Bromoimidazo[1,2-b]pyridazine 98%

Supplier: Ambeed
Warning

A123128-100G A123128-250MG A123128-25G A123128-1G A123128-5G
103800-346EA 8 USD
103800-346 77271-630 77271-632 77271-634 77312-466 77312-468
3-Bromoimidazo[1,2-b]pyridazine 98%
3-Bromoimidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C6H4BrN3
MW: 198.0201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09757672
CAS Number: 18087-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR