3-Bromoimidazo[1,2-a]pyridine 97%
Supplier: Ambeed
|
Warning
|
A481949-1G
A481949-5G
A481949-100MG
A481949-10G
A481949-250MG
103828-134EA
11
USD
103828-134
103839-208
76843-150
76843-152
77169-160
77169-162
77169-164
3-Bromoimidazo[1,2-a]pyridine 97%
3-Bromoimidazo[1,2-a]pyridine
Precaución: for Research Use Only
Formula:
C₇H₅BrN₂ Melting Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014556 CAS Number: 4926-47-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...