3-Bromoimidazo[1,2-a]pyrazine 98%

Supplier: Ambeed
Warning

A517148-1G A517148-25G A517148-5G A517148-10G A517148-250MG
103828-468EA 19 USD
103828-468 103839-574 76843-564 76843-566 76843-568 77169-482 77169-484
3-Bromoimidazo[1,2-a]pyrazine 98%
3-Bromoimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C6H4BrN3
MW: 198.0200 g/mol
Melting Pt: 191…196 °C
Storage Temperature: Ambient
MDL Number: MFCD06245372
CAS Number: 57948-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR