3-Bromofuran-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A215249-1G A215249-100MG A215249-10G A215249-250MG
103818-472EA 37 USD
103818-472 103849-176 77087-702 77275-286 77275-288 77275-290
3-Bromofuran-2-carboxylic acid 97%
3-Bromofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃BrO₃
Boiling Pt: 278 °C (760 mm Hg)
Flash Pt: 121.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00092761
CAS Number: 14903-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR