3-Bromobenzo[b]thiophene 95%

Supplier: Ambeed
Warning

A176702-25G A176702-5G A176702-100G A176702-10G A176702-1G
103807-168EA 10 USD
103807-168 103845-494 77082-846 77082-848 77191-586 77191-588 77301-468
3-Bromobenzo[b]thiophene 95%
3-Bromobenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₈H₅BrS
Boiling Pt: 269 °C (760 mm Hg)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00023009
CAS Number: 7342-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR