3-Bromobenzenesulfonyl chloride 97%

Supplier: Ambeed
Danger

A630786-25G A630786-5G A630786-10G A630786-1G
103840-272EA 9 USD
103840-272 76844-438 76844-440 77292-952 77308-008
3-Bromobenzenesulfonyl chloride 97%
3-Bromobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₄BrClO₂S
MW: 255.52 g/mol
Boiling Pt: 313.5 °C (760 mm Hg)
Melting Pt: 31…33 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052313
CAS Number: 2905-24-0
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR