3'-Bromoacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: m-Bromoacetophenone

A212968-1000G A212968-25G A212968-100G A212968-10G A212968-500G
103809-478EA 147 USD
103809-478 103848-954 77087-432 77087-434 77097-740 77097-742 77097-744
3'-Bromoacetophenone 98%
3'-Bromoacetophenone

Precaución: for Research Use Only

Formula: C₈H₇BrO
MW: 199.05 g/mol
Boiling Pt: 255.2 °C (760 mm Hg)
Melting Pt: 8…11 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000083
CAS Number: 2142-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR