3-Bromo-9H-fluoren-9-one 97%

Supplier: Ambeed
Warning

A420872-250MG A420872-5G
103827-396EA 47 USD
103827-396 77155-802 77155-804
3-Bromo-9H-fluoren-9-one 97%
3-Bromo-9H-fluoren-9-one

Precaución: for Research Use Only

Formula: C₁₃H₇BrO
Boiling Pt: 391.3±21.0 °C (760 mm Hg)
Flash Pt: 130.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00019082
CAS Number: 2041-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR