3-Bromocarbazole 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-9H-carbazole

A157959-100G A157959-10G A157959-500G A157959-1G A157959-25G A157959-5G A157959-1000G
103805-068EA 5 USD
103805-068 103843-274 76719-996 76719-998 76720-000 77300-304 77319-110 77319-112 77667-226
3-Bromocarbazole 98%
3-Bromocarbazole

Precaución: for Research Use Only

Formula: C₁₂H₈BrN
MW: 246.11 g/mol
Boiling Pt: 409.2 °C (760 mm Hg)
Melting Pt: 195 °C
Flash Pt: 201.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00222621
CAS Number: 1592-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR