3-Bromophenanthrene-9,10-dione 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-9,10-phenanthrenedione

A384435-100G A384435-10G A384435-250MG A384435-1G A384435-25G A384435-5G
103826-764EA 18 USD
103826-764 103837-802 77033-448 77033-450 77033-452 77306-952 77306-954 77306-956
3-Bromophenanthrene-9,10-dione 97%
3-Bromophenanthrene-9,10-dione

Precaución: for Research Use Only

Formula: C₁₄H₇BrO₂
MW: 287.11 g/mol
Boiling Pt: 446.8 °C (760 mm Hg)
Melting Pt: 267…268 °C
Flash Pt: 162.1 °C
Storage Temperature: Ambient
MDL Number: MFCD18072876
CAS Number: 13292-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR