3-Bromo-7-nitro-1H-indazole 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-7-nitro-1H-indazole, 3-Bromo-7-nitroindazole

A114732-100G A114732-10G A114732-250MG A114732-25G A114732-1G A114732-5G
103798-460EA 11 USD
103798-460 103824-302 77271-158 77271-160 77271-162 77271-164 77311-122 77311-124
3-Bromo-7-nitro-1H-indazole 98%
3-Bromo-7-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MW: 242.03 g/mol
Boiling Pt: 248.9 °C (760 mm Hg)
Melting Pt: 175…185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00159910
CAS Number: 74209-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR