3-Bromo-7-chloroimidazo[1,2-a]pyridine 98%
Supplier: Ambeed
|
Warning
|
A246414-100MG
A246414-250MG
A246414-25G
103811-710EA
42
USD
103811-710
103851-544
77885-638
77885-639
77885-640
3-Bromo-7-chloroimidazo[1,2-a]pyridine 98%
3-Bromo-7-chloroimidazo[1,2-a]pyridine
Precaución: Sealed in dry, at room temperature.
Formula:
C₇H₄BrClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD11111834 CAS Number: 342613-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...