3-Bromo-6-methylpyridazine 98%

Supplier: Ambeed
Warning

A113537-10G A113537-250MG A113537-25G A113537-1G A113537-5G
103798-248EA 20 USD
103798-248 103824-088 77025-888 77025-890 77025-892 77297-250 77310-904
3-Bromo-6-methylpyridazine 98%
3-Bromo-6-methylpyridazine

Precaución: for Research Use Only

Formula: C5H5BrN2
MW: 173.0106 g/mol
Boiling Pt: 285 °C (760 mm Hg)
Melting Pt: 65…69 °C
Flash Pt: 126.188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09831907
CAS Number: 65202-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR