3-Bromo-6-methoxypyridin-2-amine 97%

Supplier: Ambeed
Warning

A100356-100MG A100356-250MG A100356-1G
103795-382EA 234 USD
103795-382 103821-082 76809-194 76809-196 77296-646
3-Bromo-6-methoxypyridin-2-amine 97%
3-Bromo-6-methoxypyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MW: 203.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062946
CAS Number: 511541-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR