3-Bromo-6-fluoropyridin-2-amine 96%

Supplier: Ambeed
Warning

A153161-100MG A153161-250MG A153161-50MG A153161-5G
103804-466EA 280 USD
103804-466 76812-326 76812-328 76812-330 77883-202
3-Bromo-6-fluoropyridin-2-amine 96%
3-Bromo-6-fluoropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
Storage Temperature: Refrigerator
MDL Number: MFCD16556245
CAS Number: 1232431-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR