3-Bromo-6-fluoro-2-methylpyridine 97%
Supplier: Ambeed
|
Warning
|
A164168-100G
A164168-10G
A164168-500G
A164168-1G
A164168-25G
A164168-5G
103805-822EA
9
USD
103805-822
103844-084
77096-238
77096-240
77096-242
77300-694
77320-230
77320-232
3-Bromo-6-fluoro-2-methylpyridine 97%
3-Bromo-6-fluoro-2-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrFN Boiling Pt: 189.5 °C (760 mm Hg) Flash Pt: 68.43 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095092 CAS Number: 375368-83-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...