3-Bromo-6-chloropicolinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-6-chloropyridine-2-carboxylic acid

A266290-10G A266290-250MG A266290-1G A266290-25G A266290-5G A266290-100G
103812-794EA 12 USD
103812-794 103852-696 77031-858 77031-860 77092-394 77092-396 77092-398 77593-652
3-Bromo-6-chloropicolinic acid 97%
3-Bromo-6-chloropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MW: 236.4505 g/mol
Boiling Pt: 338.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09258777
CAS Number: 929000-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR