3-Bromo-6-chloroimidazo[1,2-b]pyridazine 97%
Supplier: Ambeed
|
Warning
|
A182799-1G
A182799-25G
A182799-5G
A182799-10G
A182799-250MG
103815-568EA
13
USD
103815-568
103846-126
77083-688
77083-690
77083-692
77151-652
77151-654
3-Bromo-6-chloroimidazo[1,2-b]pyridazine 97%
3-Bromo-6-chloroimidazo[1,2-b]pyridazine
Precaución: for Research Use Only
Formula:
C₆H₃BrClN₃ MW: 232.47 g/mol Melting Pt: 157…160 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09027274 CAS Number: 13526-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...