3-Bromo-6-chloro-5-iodopyrazin-2-amine 97%

Supplier: Ambeed
Warning

A415459-1G A415459-5G A415459-100MG A415459-250MG
103827-294EA 225 USD
103827-294 103838-338 76841-918 76841-920 77240-784 77240-786
3-Bromo-6-chloro-5-iodopyrazin-2-amine 97%
3-Bromo-6-chloro-5-iodopyrazin-2-amine

Precaución: for Research Use Only

Formula: C₄H₂BrClIN₃
MW: 334.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29472378
CAS Number: 1251941-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR