3-Bromo-6-chloro-2-fluoropyridine 95%

Supplier: Ambeed
Warning

A347000-1G A347000-25G A347000-5G A347000-10G A347000-250MG
103825-946EA 19 USD
103825-946 103836-936 76840-000 76840-002 76840-004 77278-092 77278-094
3-Bromo-6-chloro-2-fluoropyridine 95%
3-Bromo-6-chloro-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
Boiling Pt: 212 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06798230
CAS Number: 885952-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR