3-Bromo-6-chloro-2,4-dimethylpyridine 98%

Supplier: Ambeed
Warning

A129001-100MG A129001-250MG A129001-1G
103801-214EA 62 USD
103801-214 76971-960 76971-962 77298-328
3-Bromo-6-chloro-2,4-dimethylpyridine 98%
3-Bromo-6-chloro-2,4-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₇BrClN
MW: 220.49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20486518
CAS Number: 918145-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR