3-Bromo-6-chloro-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazolo[4,3-c]pyridine 95%

Supplier: Ambeed
Warning

A166997-250MG A166997-25G A166997-1G A166997-5G
103806-120EA 215 USD
103806-120 103844-382 76647-204 76647-206 77300-864 77300-866
3-Bromo-6-chloro-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazolo[4,3-c]pyridine 95%
3-Bromo-6-chloro-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazolo[4,3-c]pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₇BrClN₃OSi
Storage Temperature: Refrigerator
MDL Number: MFCD32012309
CAS Number: 2348512-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR