3-Bromo-5-nitropyridine 98%
Supplier: Ambeed
|
Danger
|
A360004-1G
A360004-25G
A360004-5G
A360004-10G
A360004-250MG
103826-242EA
27
USD
103826-242
103837-250
76840-446
76840-448
76840-450
77115-280
77115-282
3-Bromo-5-nitropyridine 98%
3-Bromo-5-nitropyridine
Precaución: for Research Use Only
Formula:
C₅H₃BrN₂O₂ Boiling Pt: 251.6 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD04114216 CAS Number: 15862-30-3 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...