2-Amino-3-bromo-5-nitropyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-nitro-2-pyridylamine, 3-Bromo-5-nitropyridin-2-amine

A102267-100G A102267-25G A102267-1G A102267-5G A102267-10G
103796-348EA 5 USD
103796-348 103822-040 76994-574 76994-576 77282-920 77282-922 77670-472
2-Amino-3-bromo-5-nitropyridine 97%
2-Amino-3-bromo-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrN₃O₂
MW: 218.01
Boiling Pt: 347.3 °C (760 mm Hg)
Melting Pt: 215…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00955628
CAS Number: 15862-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR