3-Bromo-5-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A308360-1G A308360-5G A308360-10G A308360-500G A308360-25G
103814-554EA 6 USD
103814-554 103835-730 76838-484 76838-486 77114-558 77114-560 77114-562 77305-988
3-Bromo-5-nitrobenzoic acid 97%
3-Bromo-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrNO₄
Boiling Pt: 376.8 °C (760 mm Hg)
Melting Pt: 159…161 °C
Flash Pt: 181.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00100098
CAS Number: 6307-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR