3-Bromo-5-nitrobenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A330433-1G
A330433-25G
A330433-5G
A330433-10G
A330433-250MG
103825-552EA
12
USD
103825-552
103836-512
76839-490
76839-492
76839-494
77114-874
77114-876
3-Bromo-5-nitrobenzaldehyde 97%
3-Bromo-5-nitrobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄BrNO₃ Boiling Pt: 306.9 °C (760 mm Hg) Melting Pt: 101…106 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00100097 CAS Number: 355134-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...