5-Bromo-m-cresol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-methylphenol

A186569-10G A186569-250MG A186569-1G A186569-25G A186569-5G A186569-100G
103815-944EA 6 USD
103815-944 103846-526 76985-920 76985-922 77084-190 77084-192 77084-194 77683-478
5-Bromo-m-cresol 98%
5-Bromo-m-cresol

Precaución: For research use only.

Formula: C₇H₇BrO
MW: 187.0300 g/mol
Boiling Pt: 249 °C (760 mm Hg)
Melting Pt: 61…62 °C
Flash Pt: 105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09038281
CAS Number: 74204-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR