3-Bromo-5-methylisoxazole 96%

Supplier: Ambeed
Warning

A100421-100MG A100421-250MG
103795-428EA 182 USD
103795-428 103821-124 76982-128 77682-930
3-Bromo-5-methylisoxazole 96%
3-Bromo-5-methylisoxazole

Precaución: For research use only.

Formula: C₄H₄BrNO
MW: 161.98 g/mol
Boiling Pt: 202.1 °C (760 mm Hg)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06660140
CAS Number: 25741-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR