5-Bromo-m-toluonitrile 97%

Supplier: Ambeed
Warning

A137852-100MG A137852-10G A137852-250MG A137852-1G A137852-25G A137852-5G
103802-518EA 22 USD
103802-518 77272-334 77272-336 77272-338 77315-500 77315-502 77315-504
5-Bromo-m-toluonitrile 97%
5-Bromo-m-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrN
Storage Temperature: Ambient
MDL Number: MFCD08061970
CAS Number: 124289-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR