5-Bromo-m-toluic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-5-methylbenzoic acid
A119387-100G
A119387-10G
A119387-25G
A119387-1G
A119387-5G
103799-758EA
12
USD
103799-758
76781-632
76781-634
76781-636
77297-676
77297-678
5-Bromo-m-toluic acid 98%
5-Bromo-m-toluic acid
Precaución: for Research Use Only
Formula:
C₈H₇BrO₂ MW: 215.0440 g/mol Boiling Pt: 320.3 °C (760 mm Hg) Melting Pt: 178…179.5 °C Flash Pt: 147.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD02684926 CAS Number: 58530-13-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...