3-Bromo-5-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A266875-100G A266875-10G A266875-500G A266875-1G A266875-25G A266875-5G
103812-818EA 10 USD
103812-818 103852-720 76787-098 76787-100 76787-102 77092-428 77092-430 77092-432
3-Bromo-5-methylbenzaldehyde 97%
3-Bromo-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C8H7BrO
Boiling Pt: 255.6 °C (760 mm Hg)
Flash Pt: 91.22 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09836680
CAS Number: 188813-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR