3-Bromo-5-methyl-4-nitropyridine 95%

Supplier: Ambeed
Warning

A198646-250MG A198646-1G A198646-5G
103817-038EA 282 USD
103817-038 103847-666 77012-584 77302-598 77302-600
3-Bromo-5-methyl-4-nitropyridine 95%
3-Bromo-5-methyl-4-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17014982
CAS Number: 1379323-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR