3-Bromo-5-methyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

A183809-100G A183809-10G A183809-1G A183809-25G A183809-5G
103815-674EA 8 USD
103815-674 103846-236 77029-548 77029-550 77083-836 77083-838 77083-840
3-Bromo-5-methyl-1H-pyrazole 98%
3-Bromo-5-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C4H5BrN2
Boiling Pt: 270 °C (760 mm Hg)
Flash Pt: 117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11215487
CAS Number: 57097-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR