3-Bromo-5-methoxypyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-3-methoxypyridine
A217924-1000G
A217924-100G
A217924-10G
A217924-500G
A217924-25G
A217924-5G
103849-400EA
7
USD
103849-400
76975-746
76975-748
76975-750
76975-752
77088-042
77088-044
3-Bromo-5-methoxypyridine 98%
3-Bromo-5-methoxypyridine
Precaución: for Research Use Only
Formula:
C₆H₆BrNO MW: 188.02 g/mol Boiling Pt: 212.2 °C (760 mm Hg) Melting Pt: 31…35 °C Flash Pt: 102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234169 CAS Number: 50720-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...