3-Bromo-5-methoxybenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A278482-1G
A278482-25G
A278482-5G
A278482-10G
A278482-250MG
103813-414EA
7
USD
103813-414
103853-356
77093-278
77093-280
77093-282
77276-912
77276-914
3-Bromo-5-methoxybenzonitrile 98%
3-Bromo-5-methoxybenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₆BrNO MW: 212.05 g/mol Boiling Pt: 253.5 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08061971 CAS Number: 867366-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...