3-Bromo-5-methoxybenzoic acid 97%

Supplier: Ambeed
Warning

A211742-100G A211742-10G A211742-250MG A211742-1G A211742-25G A211742-5G
103818-134EA 16 USD
103818-134 103848-830 76785-692 76785-694 76785-696 77087-252 77087-254 77087-256
3-Bromo-5-methoxybenzoic acid 97%
3-Bromo-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
Storage Temperature: Ambient
MDL Number: MFCD00266858
CAS Number: 157893-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR